missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NXT2 Protein

A cDNA sequence encoding the NXT2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12914-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941939

  • £155.00 / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NXT2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMRKYRS HWSQGDREGY QRRSNYYEGP HTSHSSPADR TREEVVTPTL PEHTATRSQM ATSLDFKTYV DQACRAAEEF VNIYYETMDK RRRALTRLYL DKATLIWNGN AVSGLDALNN FFDTLPSSEF QVNMLDCQPV HEQATQSQTT VLVVTSGTVKFDGNKQHFFN QNFLLTAQST PNNTVWKIAS DCFRFQDWSS S
Greater than 95.0% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NXT2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 40% glycerol and 200mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers