missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDT16 Protein
A cDNA sequence encoding the NUDT16 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12902-5ug
This item is not returnable.
View return policy
Specifications
NUDT16 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAGARRLELG EALALGSGWR HACHALLYAP DPGMLFGRIP LRYAILMQMR FDGRLGFPGG FVDTQDRSLE DGLNRELREE LGEAAAAFRV ERTDYRSSHV GSGPRVVAHF YAKRLTLEEL LAVEAGATRA KDHGLEVLGL VRVPLYTLRD GVGGLPTFLE NSFIGSAREQ LLEALQDLGL LQSGSISGLK IPAHH | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NUDT16 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 100mM NaCl and 10% glycerol. |