missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleoside diphosphate kinase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Nucleoside diphosphate kinase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6425-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273362.1 | |
Nucleoside diphosphate kinase, mitochondrial Protein | |
Research Use Only | |
NME4 | |
Recombinant Protein | |
E. coli | |
SWTRERTLVA VKPDGVQRRL VGDVIQRFER RGFTLVGMKM LQAPESVLAE HYQDLRRKPF YPALIRYMSS GPVVAMVWEG YNVVRASRAM IGHTDSAEAA PGTIRGDFSV HISRNVIHAS DSVEGAQREI QLWFQSSELV SWADGGQHSS IHPA | |
Greater than 80% as determined by SDS-PAGE. |
4833 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |