missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolar protein 1
A DNA sequence encoding the Homo sapiens (Human) Nucleolar protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6427-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001171986.1 | |
Nucleolar protein 1 | |
Research Use Only | |
NOL3 | |
Recombinant Protein | |
E. coli | |
MGNAQERPSE TIDRERKRLV ETLQADSGLL LDALLARGVL TGPEYEALDA LPDAERRVRR LLLLVQGKGE AACQELLRCA QRTAGAPDPA WDWQHVGPGY RDRSYDPPCP GHWTPEAPGS GTTCPGLPRA SDPDEAGGPE GSEAVQSGTP EEPEPELEAE ASKEAEPEPE PEPELEPEAE AEPEPELEPE PDPEPEPDFE ERDESEDS | |
Greater than 90% as determined by SDS-PAGE. |
8996 | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |