missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NTF4 Protein
Recombinant Protein
Brand: enQuireBio™ QP5345-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_006170.1 | |
NTF4 Protein | |
Research Use Only | |
NTF4 | |
Recombinant Protein | |
E. coli | |
MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A | |
Greater than 95% as determined by SDS-PAGE. |
4909 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human NTF4 induces Rat C6 cell proliferation. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |