missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human NT-proBNP Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
Data is not available.
Specifications
Specifications
| For Use With (Application) | Western Blot, SDS-PAGE |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM Tris-HCl, pH 8.0, 150 mM NaCl. |
| Gene ID (Entrez) | 4879 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 8.5 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of NT-proBNP as determined by LAL method. |
| Gene Alias | BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein |
| Gene Symbol | NPPB |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction