missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human NT-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£200.86 - £530.22
Specifications
Gene ID (Entrez) | 4909 |
---|---|
Molecular Weight (g/mol) | 14 kDa |
Name | NT-4 |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human NT-4 protein Gly81-Ala210 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18176430
|
R&D Systems™
268-N4-005 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 5μg |
£200.86
5µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18310278
|
R&D Systems™
268-N4-005/CF |
£200.86
5µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18127880
|
R&D Systems™
268-N4-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 25μg |
£530.22
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18147140
|
R&D Systems™
268-N4-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | 25μg |
£530.22
25µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Recombinant Human NT-4 Protein is derived from Sf 21 (baculovirus). The Recombinant Human NT-4 Protein has been validated for the following applications: Bioactivity.Specifications
4909 | |
NT-4 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
14 kDa | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human NT-4 protein Gly81-Ala210 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |