missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NPM2 Protein
A cDNA sequence encoding the NPM2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12872-1mg
Additional Details : Weight : 0.01000kg
Specifications
NPM2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNLSSAS STEEKAVTTV LWGCELSQER RTWTFRPQLE GKQSCRLLLH TICLGEKAKE EMHRVEILPP ANQEDKKMQP VTIASLQASV LPMVSMVGVQ LSPPVTFQLR AGSGPVFLSG QERYEASDLT WEEEEEEEGE EEEEEEEDDE DEDADISLEE QSPVKQVKRL VPQKQASVAK KKKLEKEEEE IRASVRDKSP VKKAKATARA KKPGFKK | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NPM2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol. |