missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCN3 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1.0 μg/mL, corresponding to a specific activity of >1000 IU/mg.
Specifications
Specifications
| Accession Number | P48745, P48745, P48745 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris-HCl, pH 8.6, 150 mM NaCl. |
| Gene ID (Entrez) | 4856 |
| Molecular Weight (g/mol) | 36.2 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of NOV/CCN3 as determined by LAL method. |
| Gene Alias | CCN family member 3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?