missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Notch-4 Fc Chimera Protein

Product Code. 18329417
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329417 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329417 Supplier R&D Systems™ Supplier No. 9909NT050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

  • Derived from T. ni (baculovirus)
  • Validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99466-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 4855
Molecular Weight (g/mol) 94 to 116 kDa, reducing conditions
Quantity 50 μg
Source Trichoplusia ni, T. ni (baculovirus)-derived human Notch-4 protein. Human Notch-4 (Arg24-Glu549) Accession # Q99466-1IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 400 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Purity or Quality Grade > 90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.