missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Non-secretory ribonuclease Protein

A DNA sequence encoding the Homo sapiens (Human) Non-secretory ribonuclease, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6610-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084520

  • £1735.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002925.1
Non-secretory ribonuclease Protein
Research Use Only
RNASE2
Recombinant Protein
E. coli
KPPQFTWAQW FETQHINMTS QQCTNAMQVI NNYQRRCKNQ NTFLLTTFAN VVNVCGNPNM TCPSNKTRKN CHHSGSQVPL IHCNLTTPSP QNISNCRYAQ TPANMFYIVA CDNRDQRRDP PQYPVVPVHL DRII
Greater than 90% as determined by SDS-PAGE.
6036
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers