missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Noggin Fc Chimera Protein
Description
Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.100-0.400 µg/mL in the presence of 30 ng/mL of Recombinant Human BMP-4 (Catalog # 314-BP ).
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 9241 |
| Molecular Weight (g/mol) | 49.6 kDa (monomer) |
| Name | Noggin Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Noggin protein, (N-terminus) Human Noggin (Gln28-Cys232) Accession &Num; Q13253 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?