missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human NKp80/KLRF1 Fc Chimera Protein

Product Code. 18368767
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368767 50 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368767 Supplier R&D Systems™ Supplier No. 1900NK050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human NKp80/KLRF1 Fc Chimera Protein is derived from NS0. The Recombinant Human NKp80/KLRF1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 51348
Molecular Weight (g/mol) 45.8 kDa (monomer)
Name NKp80/KLRF1 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human NKp80/KLRF1 protein, (N-terminus) MD Human IgG1 (Pro100-Lys330) IEGR Human NKp80 (Val66-Tyr231) Accession &Num; Q9NZS2 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.