missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NKIRAS2 Protein

A cDNA sequence encoding the NKIRAS2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12849-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951739

  • £155.00 / 10µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NKIRAS2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMGKSCK VVVCGQASVG KTSILEQLLY GNHVVGSEMI ETQEDIYVGS IETDRGVREQ VRFYDTRGLR DGAELPRHCF SCTDGYVLVY STDSRESFQR VELLKKEIDK SKDKKEVTIV VLGNKCDLQE QRRVDPDVAQ HWAKSEKVKL WEVSVADRRSLLEPFVYLAS KMTQPQSKSA FPLSRKNKGS GSLDG
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NKIRAS2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers