missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NIP7 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) NIP7 homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8287-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186363.1 | |
NIP7 homolog Protein | |
Research Use Only | |
NIP7 | |
Recombinant Protein | |
E. coli | |
MRPLTEEETR VMFEKIAKYI GENLQLLVDR PDGTYCFRLH NDRVYYVSEK IMKLAANISG DKLVSLGTCF GKFTKTHKFR LHVTALDYLA PYAKYKVWIK PGAEQSFLYG NHVLKSGLGR ITENTSQYQG VVVYSMADIP LGFGVAAKST QDCRKVDPMA IVVFHQADIG EYVRHEETLT | |
Greater than 90% as determined by SDS-PAGE. |
51388 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |