missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Ninjurin-2 Protein

Product Code. 18390548
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390548 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390548 Supplier R&D Systems™ Supplier No. 9719NJ050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Derived from E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZG7
For Use With (Application) Flow Cytometry
Gene ID (Entrez) 4815
Molecular Weight (g/mol) 8 kDa, reducing conditions
Quantity 50 μg
Source E. coli-derived human Ninjurin-2 protein Met1-Thr65, wtih a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Purity or Quality Grade > 95%, by SDS-PAGE under reducing conditions and visualized by silver Stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.