missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Nicotinic Acetylcholine Receptor beta 2 GST (N-Term) Protein

Product Code. 18380158 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380158 10 μg 10µg
18304298 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18380158 Supplier Novus Biologicals™ Supplier No. H00001141Q0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 1141
Molecular Weight (g/mol) M.W. (Theoretical): 37.29 kDa
Quantity 10 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias cholinergic receptor, nicotinic, beta 2 (neuronal), cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal), EFNL3, nAChRB2, neuronal acetylcholine receptor subunit beta-2, neuronal nicotinic acetylcholine receptor beta 2
Gene Symbol CHRNB2
Research Category Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Nicotinic Acetylcholine Receptor beta 2
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.