missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Neurturin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
£200.86
Specifications
Gene ID (Entrez) | 4902 |
---|---|
Molecular Weight (g/mol) | 11.8 kDa (monomer) |
Name | Neurturin |
Quantity | 25μg |
Source | E. coli-derived human Neurturin protein Ala96-Val197,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18158300
|
R&D Systems™
1297-NE-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£200.86
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18146980
|
R&D Systems™
1297-NE-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£200.86
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human Neurturin Protein is derived from E. coli. The Recombinant Human Neurturin Protein has been validated for the following applications: Bioactivity, Binding Activity.Specifications
4902 | |
Neurturin | |
E. coli-derived human Neurturin protein Ala96-Val197,with an N-terminal Met | |
Disulfide-linked homodimer | |
Recombinant |
11.8 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |