missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Neurexophilin-2 Protein

Product Code. 15912848
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 15912848

Brand: enQuireBio™ QP10093ma20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Neurexophilin-2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_009157.1
Gene ID (Entrez) 11249
Name Neurexophilin-2 Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol NXPH2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Mammal
Protein Tag His
Sequence KEVVHATEGL DWEDKDAPGT LVGNVVHSRI ISPLRLFVKQ SPVPKPGPMA YADSMENFWD WLANITEIQE PLARTKRRPI VKTGKFKKMF GWGDFHSNIK TVKLNLLITG KIVDHGNGTF SVYFRHNSTG LGNVSVSLVP PSKVVEFEVS PQSTLETKES KSFNCRIEYE KTDRAKKTAL CNFDPSKICY QEQTQSHVSW LCSKPFKVIC IYIAFYSVDY KLVQKVCPDY NYHSETPYLS SG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.