missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Neuregulin-1 alpha/NRG1 alpha Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
missing translation for 'unitSize'
1000µg
100µg
10µg
500µg
Description
Neuregulin-1 alpha/NRG1 alpha protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using serum free human MCF-7 cells is less than 40 ng/mL, corresponding to a specific activity of >2.5 x 10^4 IU/mg.
Specifications
Specifications
| Accession Number | Q02297, Q02297, Q02297 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in 20 mM PB, pH 6.0, 150 mM NaCl. |
| Gene ID (Entrez) | 3084 |
| Molecular Weight (g/mol) | 7.4 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Neuregulin-1 alpha/NRG1 alpha as determined by LAL method. |
| Gene Alias | ARIA, GGF2, HGL, HRG, HRG1, MST131, MSTP131, neuregulin 1, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction