missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NENF Protein

A cDNA sequence encoding the NENF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12829-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951679

  • £155.00 / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NENF Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASGQTPRPAERG PPVRLFTEEE LARYGGEEED QPIYLAVKGV VFDVTSGKEF YGRGAPYNAL TGKDSTRGVA KMSLDPADLT HDTTGLTAKE LEALDEVFTK VYKAKYPIVG YTARRILNED GSPNLDFKPE DQPHFDIKDE F
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NENF was filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers