missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NEIL2 Protein
A cDNA sequence encoding the NEIL2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12826-1mg
Additional Details : Weight : 0.01000kg
Specifications
252969 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMPEGPL VRKFHHLVSP FVGQQVVKTG GSSKKLQPAS LQSLWLQDTQ VHGKKLFLRF DLDEEMGPPG SSPTPEPPQK EVQKEGAADP KQVGEPSGQK TLDGSSRSAE LVPQGEDDSE YLERDAPAGD AGRWLRVSFG LFGSVWVNDF SRAKKANKRG DWRDPSPRLV LHFGGGGFLA FYNCQLSWSS SPVVTPTCDI LSEKFHRGQA LEALGQAQPV CYTLLDQRYF SGLGNIIKNE ALYRAGIHPL SLGSVLSASR REVLVDHVVE FSTAWLQGKF QGRPQHTQVY QKEQCPAGHQ VMKEAFGPED GLQRLTWWCP QCQPQLSEEP EQCQFS | |
Greater than 85.0% as determined by SDS-PAGE. |
NEIL2 Protein | |
Research Use Only | |
NEIL2 | |
Human | |
His | |
The NEIL2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |