missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NEDD8 Protein

A DNA sequence encoding the Homo sapiens (Human) NEDD8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7554-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049350

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006147.1
NEDD8 Protein
Research Use Only
NEDD8
Recombinant Protein
E. coli
MLIKVKTLTG KEIEIDIEPT DKVERIKERV EEKEGIPPQQ QRLIYSGKQM NDEKTAADYK ILGGSVLHLV LALRGG
Greater than 80% as determined by SDS-PAGE.
4738
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers