missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Nectin-2/CD112 Protein

Product Code. 18177570
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in PBS.
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18177570

Brand: R&D Systems™ 2229N2050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to bind with Recombinant Human DNAM-1/CD226 Fc Chimera (Catalog # 666-DN ) in a functional ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_002847
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 5819
Molecular Weight (g/mol) M.W. (Observed): 43-50 kDa, reducing conditions; M.W. (theoretical): 36.3 kDa
Name Nectin-2/CD112
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Nectin-2/CD112 protein Gln32-Leu360, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Monomer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol NECTIN2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Nectin-2/CD112
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.