missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFV2 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFV2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8505-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_066552.2 | |
NDUFV2 Protein | |
Research Use Only | |
NDUFV2 | |
Recombinant Protein | |
E. coli | |
GGALFVHRDT PENNPDTPFD FTPENYKRIE AIVKNYPEGH KAAAVLPVLD LAQRQNGWLP ISAMNKVAEV LQVPPMRVYE VATFYTMYNR KPVGKYHIQV CTTTPCMLRN SDSILEAIQK KLGIKVGETT PDKLFTLIEV ECLGACVNAP MVQINDNYYE DLTAKDIEEI IDELKAGKIP KPGPRSGRFS CEPAGGLTSL TEPPKGPGFG VQAGL | |
Greater than 90% as determined by SDS-PAGE. |
4729 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |