missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFB4 Protein
A cDNA sequence encoding the NDUFB4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12816-5ug
Additional Details : Weight : 0.01000kg
Specifications
4710 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSFPKYK PSSLRTLPET LDPAEYNISP ETRRAQAERL AIRAQLKREY LLQYNDPNRR GLIENPALLR WAYARTINVY PNFRPTPKNS | |
Greater than 90% as determined by SDS-PAGE. |
NDUFB4 Protein | |
Research Use Only | |
NDUFB4 | |
Human | |
His | |
The NDUFB4 solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea And 10% glycerol. |