missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFB4 Protein

A cDNA sequence encoding the NDUFB4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12816-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961639

  • £2975.00 / 1mg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4710
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSFPKYK PSSLRTLPET LDPAEYNISP ETRRAQAERL AIRAQLKREY LLQYNDPNRR GLIENPALLR WAYARTINVY PNFRPTPKNS
Greater than 90% as determined by SDS-PAGE.
NDUFB4 Protein
Research Use Only
NDUFB4
Human
His
The NDUFB4 solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea And 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers