missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFA6 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFA6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6411-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002481.2 | |
NDUFA6 Protein | |
Research Use Only | |
NDUFA6 | |
Recombinant Protein | |
E. coli | |
MAGSGVRQAT STASTFVKPI FSRDMNEAKR RVRELYRAWY REVPNTVHQF QLDITVKMGR DKVREMFMKN AHVTDPRVVD LLVIKGKIEL EETIKVWKQR THVMRFFHET EAPRPKDFLS KFYVGHDP | |
Greater than 90% as determined by SDS-PAGE. |
4700 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |