missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFA6 Protein

A DNA sequence encoding the Homo sapiens (Human) NDUFA6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6411-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023120

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002481.2
NDUFA6 Protein
Research Use Only
NDUFA6
Recombinant Protein
E. coli
MAGSGVRQAT STASTFVKPI FSRDMNEAKR RVRELYRAWY REVPNTVHQF QLDITVKMGR DKVREMFMKN AHVTDPRVVD LLVIKGKIEL EETIKVWKQR THVMRFFHET EAPRPKDFLS KFYVGHDP
Greater than 90% as determined by SDS-PAGE.
4700
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers