missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human NDST2 Protein
Description
Measured by its ability to transfer sulfate from PAPS to heparan sulfate. The specific activity is >1,250 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P52849 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 8509 |
| Molecular Weight (g/mol) | M.W. (Observed): 90-100 kDa, reducing conditions; M.W. (theoretical): 94 kDa |
| Name | N-Deacetylase/N-Sulfotransferase 2/NDST2 |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell, HEK293-derived human N-Deacetylase/N-Sulfotransferase 2/NDST2 protein Ala81-Gly883, with C-terminal addition of HPGGGSGGGSGGGSHHHHHH |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?