missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDST1 Protein

Product Code. 16093100
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16093100

missing translation for 'mfr': enQuireBio™ QP6409ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) NDST1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001287992.1
Gene ID (Entrez) 3340
Name NDST1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol NDST1
Protein Length Full Length of Isoform 2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence GWKRGLEPSA DAPEPDCGDP PPVAPSRLLP LKPVQAATPS RTDPLVLVFV ESLYSQLGQE VVAILESSRF KYRTEIAPGK GDMPTLTDKG RGRFALIIYE NILKYVNLDA WNRELLDKYC VAYGVGIIGF FKANENSLLS AQLKGFPLFL HSNLGLKDCS INPKSPLLYV TRPSEVEKGV LPGEDWTVFQ SNHSTYEPVL LAKTRSSESI PHLGADAGLH AALHATVVQD LGLHDGIQRV LFGNNLNFWL HKLVFVDAVA FLTGKRLSLP LDRYILVDID DIFVGKEGTR MKVEDVKALF DTQNELRAHI PNFTFNLGYS GKFFHTGTNA EDAGDDLLLS YVKEFWWFPH MWSHMQPHLF HNQSVLAEQM ALNKKFAVEH GIPTDMGYAV APHHSGVYPV HVQLYEAWKQ VWSIRVTSTE EYPHLKPARY RRGFIHNGIM VLPRQTCGLF THTIFYNEYP GGSSELDKII NGGELFLTVL LNPVSAPQPM AAGEKGLLHS LSAADTGFLE PGKGGEA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 83.3% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.