missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDST1 Protein
A DNA sequence encoding the Homo sapiens (Human) NDST1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6409-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001287992.1 | |
NDST1 Protein | |
Research Use Only | |
NDST1 | |
Recombinant Protein | |
E. coli | |
GWKRGLEPSA DAPEPDCGDP PPVAPSRLLP LKPVQAATPS RTDPLVLVFV ESLYSQLGQE VVAILESSRF KYRTEIAPGK GDMPTLTDKG RGRFALIIYE NILKYVNLDA WNRELLDKYC VAYGVGIIGF FKANENSLLS AQLKGFPLFL HSNLGLKDCS INPKSPLLYV TRPSEVEKGV LPGEDWTVFQ SNHSTYEPVL LAKTRSSESI PHLGADAGLH AALHATVVQD LGLHDGIQRV LFGNNLNFWL HKLVFVDAVA FLTGKRLSLP LDRYILVDID DIFVGKEGTR MKVEDVKALF DTQNELRAHI PNFTFNLGYS GKFFHTGTNA EDAGDDLLLS YVKEFWWFPH MWSHMQPHLF HNQSVLAEQM ALNKKFAVEH GIPTDMGYAV APHHSGVYPV HVQLYEAWKQ VWSIRVTSTE EYPHLKPARY RRGFIHNGIM VLPRQTCGLF THTIFYNEYP GGSSELDKII NGGELFLTVL LNPVSAPQPM AAGEKGLLHS LSAADTGFLE PGKGGEA | |
Greater than 83.3% as determined by SDS-PAGE. |
3340 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |