missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDRG2 Protein
A cDNA sequence encoding the NDRG2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12809-20ug
Weitere Details : Weight : 0.01000kg
Spezifikation
NDRG2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAELQE VQITEEKPLL PGQTPEAAKT HSVETPYGSV TFTVYGTPKP KRPAILTYHD VGLNYKSCFQ PLFQFEDMQE IIQNFVRVHV DAPGMEEGAP VFPLGYQYPS LDQLADMIPC VLQYLNFSTI IGVGVGAGAY ILARYALNHP DTVEGLVLIN IDPNAKGWMD WAAHKLTGLT SSIPEMILGH LFSQEELSGN SELIQKYRNI ITHAPNLDNI ELYWNSYNNR RDLNFERGGD ITLRCPVMLV VGDQAPHEDA VVECNSKLDP TQTSFLKMAD SGGQPQLTQP GKLTEAFKYF LQGMGYMASS CMTRLSRSRT ASLTSAASVD GNRSRSRTLS QSSESGTLSS GPPGHTMEVS C | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NDRG2 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 0.4M Urea and 10% glycerol. |