missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDFIP1 Protein
A cDNA sequence encoding the NDFIP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12806-5ug
Additional Details : Weight : 0.01000kg
Specifications
NDFIP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMALALAA LAAVEPACGS RYQQLQNEEE SGEPEQAAGD APPPYSSISA ESAAYFDYKD ESGFPKPPSY NVATTLPSYD EAERTKAEAT IPLVPGRDED FVGRDDFDDA DQLRIGNDG | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NDFIP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |