missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDFIP1 Protein

A cDNA sequence encoding the NDFIP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12806-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941599

  • £2975.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NDFIP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMALALAA LAAVEPACGS RYQQLQNEEE SGEPEQAAGD APPPYSSISA ESAAYFDYKD ESGFPKPPSY NVATTLPSYD EAERTKAEAT IPLVPGRDED FVGRDDFDDA DQLRIGNDG
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NDFIP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers