missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Napsin A Protein
Description
The Recombinant Human Napsin A Protein is derived from CHO. The Recombinant Human Napsin A Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Accession Number | O96009 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 9476 |
| Molecular Weight (g/mol) | M.W. (Observed): 40-70 kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Napsin A protein Thr25-Gly420,with C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU per 1μg of the protein by the LAL method. |
| Gene Alias | Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPA, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?