missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human NAGLU Protein
Click to view available options
:
20ug; Carrier Free
Description
Measured by its ability to hydrolyze 4-Nitrophenyl-N-acetyl-alpha-D-glucosaminide. The specific activity is >900 pmol/min/μg, as measured under the decribed conditions.
Specifications
Specifications
| Accession Number | P54802 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 4669 |
| Molecular Weight (g/mol) | M.W. (Observed): 70-95 kDa, reducing conditions; M.W. (theoretical): 81 kDa |
| Name | alpha-N-acetylglucosaminidase/NAGLU |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human alpha-N-acetylglucosaminidase/NAGLU protein Met1-Trp743, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction