missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NAE1 Protein

A cDNA sequence encoding the NAE1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12783-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941529

  • £99.15 / 2µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NAE1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAQLGKL LKEQKYDRQL RLWGDHGQEA LESAHVCLIN ATATGTEILK NLVLPGIGSF TIIDGNQVSG EDAGNNFFLQ RSSIGKNRAE AAMEFLQELN SDVSGSFVEE SPENLLDNDP SFFCRFTVVV ATQLPESTSL RLADVLWNSQ IPLLICRTYG LVGYMRIIIK EHPVIESHPD NALEDLRLDK PFPELREHFQ SYDLDHMEKK DHSHTPWIVI IAKYLAQWYS ETNGRIPKTY KEKEDFRDLI RQGILKNENG APEDEENFEE AIKNVNTALN TTQIPSSIED IFNDDRCINI TKQTPSFWIL ARALKEFVAK EGQGNLPVRG TIPDMIADSG KYIKLQNVYR EKAKKDAAAV GNHVAKLLQS IGQAPESISE KELKLLCSNS AFLRVVRCRS LAEEYGLDTI NKDEIISSMD NPDNEIVLYL MLRAVDRFHK QQGRYPGVSN YQVEEDIGKL KSCLTGFLQE YGLSVMVKDD YVHEFCRYGA AEPHTIAAFL GGAAAQEVIK IITKQFVIFN NTYIYSGMSQ TSATFQL
Greater than 90% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NAE1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 200mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers