missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAA30 Protein
A cDNA sequence encoding the NAA30 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12781-2ug
Additional Details : Weight : 0.01000kg
Specifications
122830 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEVPPG PSSLLPPPAP PAPAAVEPRC PFPAGAALAC CSEDEEDDEE HEGGGSRSPA GGESATVAAK GHPCLRCPQP PQEQQQLNGL ISPELRHLRA AASLKSKVLS VAEVAATTAT PDGGPRATAT KGAGVHSGER PPHSLSSNAR TAVPSPVEAA AASDPAAARN GLAEGTEQEE EEEDEQVRLL SSSLTADCSL RSPSGREVEP GEDRTIRYVR YESELQMPDI MRLITKDLSE PYSIYTYRYF IHNWPQLCFL AMVGEECVGA IVCKLDMHKK MFRRGYIAML AVDSKYRRNG IGTNLVKKAI YAMVEGDCDE VVLETEITNK SALKLYENLG FVRDKRLFRY YLNGVDALRL KLWLR | |
Greater than 90.0% as determined by SDS-PAGE. |
NAA30 Protein | |
Research Use Only | |
NAA30 | |
Human | |
His | |
NAA30 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |