missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N-acetyl-D-glucosamine kinase Protein
A DNA sequence encoding the Homo sapiens (Human) N-acetyl-D-glucosamine kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8192-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060037.3 | |
N-acetyl-D-glucosamine kinase Protein | |
Research Use Only | |
NAGK | |
Recombinant Protein | |
E. coli | |
AAIYGGVEGG GTRSEVLLVS EDGKILAEAD GLSTNHWLIG TDKCVERINE MVNRAKRKAG VDPLVPLRSL GLSLSGGDQE DAGRILIEEL RDRFPYLSES YLITTDAAGS IATATPDGGV VLISGTGSNC RLINPDGSES GCGGWGHMMG DEGSAYWIAH QAVKIVFDSI DNLEAAPHDI GYVKQAMFHY FQVPDRLGIL THLYRDFDKC RFAGFCRKIA EGAQQGDPLS RYIFRKAGEM LGRHIVAVLP EIDPVLFQGK IGLPILCVGS VWKSWELLKE GFLLALTQGR EIQAQNFFSS FTLMKLRHSS ALGGASLGAR HIGHLLPMDY SANAIAFYSY TFS | |
Greater than 90% as determined by SDS-PAGE. |
55577 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |