missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human N-acetyl-D-glucosamine kinase Protein

A DNA sequence encoding the Homo sapiens (Human) N-acetyl-D-glucosamine kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8192-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013181

  • £1735.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_060037.3
N-acetyl-D-glucosamine kinase Protein
Research Use Only
NAGK
Recombinant Protein
E. coli
AAIYGGVEGG GTRSEVLLVS EDGKILAEAD GLSTNHWLIG TDKCVERINE MVNRAKRKAG VDPLVPLRSL GLSLSGGDQE DAGRILIEEL RDRFPYLSES YLITTDAAGS IATATPDGGV VLISGTGSNC RLINPDGSES GCGGWGHMMG DEGSAYWIAH QAVKIVFDSI DNLEAAPHDI GYVKQAMFHY FQVPDRLGIL THLYRDFDKC RFAGFCRKIA EGAQQGDPLS RYIFRKAGEM LGRHIVAVLP EIDPVLFQGK IGLPILCVGS VWKSWELLKE GFLLALTQGR EIQAQNFFSS FTLMKLRHSS ALGGASLGAR HIGHLLPMDY SANAIAFYSY TFS
Greater than 90% as determined by SDS-PAGE.
55577
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers