missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myoglobin Protein

Product Code. 15981499
Click to view available options
Quantity:
1 mg
10 μg
10 mg
200 μg
50 μg
Unit Size:
10mg
10µg
1mg
200µg
50µg
This item is not returnable. View return policy

Product Code. 15981499

Brand: enQuireBio™ QP1277610mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Myoglobin was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 4151
Name Myoglobin Protein
Quantity 10 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol MB
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Sequence MGLSDGEWQL VLNVWGKVEA DIPGHGQEVL IRLFKGHPET LEKFDKFKHL KSEDEMKASE DLKKHGATVL TALGGILKKK GHHEAEIKPL AQSHATKHKI PVKYLEFISE CIIQVLQSKH PGDFGADAQG AMNKALELFR KDMASNYKEL GFQG
Buffer The protein solution is in 0.05M phosphate buffer pH 7.5 containing 0.15M NaCl and 0.09% NaN3. Filtered through a 0.2microm membrane.
Purity or Quality Grade Greater than 96.0%.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.