missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MYL12A Protein

A cDNA sequence encoding the MYL12A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12767-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931479

  • £155.00 / 20µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MYL12A Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSSKRT KTKTKKRPQR ATSNVFAMFD QSQIQEFKEA FNMIDQNRDG FIDKEDLHDM LASLGKNPTD EYLDAMMNEA PGPINFTMFL TMFGEKLNGT DPEDVIRNAF ACFDEEATGT IQEDYLRELL TTMGDRFTDE EVDELYREAP IDKKGNFNYI EFTRILKHGA KDKDD
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MYL12A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers