missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYL12A Protein
A cDNA sequence encoding the MYL12A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12767-1mg
Additional Details : Weight : 0.01000kg
Specifications
MYL12A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSSKRT KTKTKKRPQR ATSNVFAMFD QSQIQEFKEA FNMIDQNRDG FIDKEDLHDM LASLGKNPTD EYLDAMMNEA PGPINFTMFL TMFGEKLNGT DPEDVIRNAF ACFDEEATGT IQEDYLRELL TTMGDRFTDE EVDELYREAP IDKKGNFNYI EFTRILKHGA KDKDD | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MYL12A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |