missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 4611 |
| Molecular Weight (g/mol) | 36.85 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | bHLHe38, L-MYC2, MYCL1P1, MYCL2, MYCL-related processed, v-myc avian myelocytomatosis viral oncogene homolog 1 pseudogene 1, v-myc avian myelocytomatosis viral oncogene homolog 2, v-myc myelocytomatosis viral oncogene homolog 1 pseudogene 1, v-myc myelocytomatosis viral oncogene homolog 2, v-myc myelocytomatosis viral oncogene homolog 2 (avian) |
| Gene Symbol | MYCL2 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | MYCL2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?