missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human MuSK Fc Chimera Protein

Product Code. 18711399
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18711399

Brand: R&D Systems™ 9810MK050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. In the presence of Recombinant Human Agrin (Catalog # 6624-AG ), immobilized Recombinant Human MuSK Fc Chimera (2 μg/mL at 100 μL/well) binds Recombinant Human LRP-4 (Catalog # 5948-LR ) with an ED50 of 5-50 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15146-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 4593
Molecular Weight (g/mol) 78 kDa
Name EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human MuSK protein, (N-terminus) Human MuSK (Leu24-Thr495) Accession &Num; O15146-1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.