missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mucosal addressin cell adhesion molecule 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7559-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049380

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_570116.2
Mucosal addressin cell adhesion molecule 1 Protein
Research Use Only
Madcam1
Recombinant Protein
E. coli
QSLQVKPLQV EPPEPVVAVA LGASRQLTCR LACADRGASV QWRGLDTSLG AVQSDTGRSV LTVRNASLSA AGTRVCVGSC GGRTFQHTVQ LLVYAFPDQL TVSPAALVPG DPEVACTAHK VTPVDPNALS FSLLVGGQEL EGAQALGPEV QEEEEEPQGD EDVLFRVTER WRLPPLGTPV PPALYCQATM RLPGLELSHR QAIPVLHSPT SPEPPDTTSP ESPDTTSPES PDTTSQEPPD TTSPEPPDKT SPEPAPQQGS THTPRSPGST RTRRPEISQA GPTQGEVIPT GSSKPAGDQ
Greater than 80% as determined by SDS-PAGE.
8174
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers