missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MSR1 Protein

A cDNA sequence encoding the MSR1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12740-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951389

  • £155.00 / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4481
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSKWETKNC SVSSTNANDI TQSLTGKGND SEEEMRFQEV FMEHMSNMEK RIQHILDMEA NLMDTEHFQN FSMTTDQRFN DILLQLSTLF SSVQGHGNAI DEISKSLISL NTTLLDLQLN IENLNGKIQE NTFKQQEEIS KLEERVYNVS AEIMAMKEEQ VHLEQEIKGE VKVLNNITND LRLKDWEHSQ TLRNITLIQG PPGPPGEKGD RGPTGESGPR GFPGPIGPPG LKGDRGAIGF PGSRGLPGYA GRPGNSGPKG QKGEKGSGNT LTPFTKVRLV GGSGPHEGRV EILHSGQWGT ICDDRWEVRV GQVVCRSLGY PGVQAVHKAA HFGQGTGPIW LNEVFCFGRE SSIEECKIRQ WGTRACSHSE DAGVTCTL
Greater than 85.0% as determined by SDS-PAGE.
MSR1 Protein
Research Use Only
MSR1
Human
His
MSR1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers