missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human MSP/MST1 Protein

Product Code. 18146350
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18146350

Brand: R&D Systems™ 352MS050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to activate MSP R/Ron in MDA-MB-453 or T47D human breast cancer cells. 80 ng/mL of the recombinant human MSP significantly induces phosphorylation of MSP R/Ron measured by DuoSet IC human phospho-MSP R/Ron kit (Catalog # DYC1947 ). Measured by its binding ability in a functional ELISA. When Recombinant Human MSPR/Ron Protein (Catalog #1947-MS) is represent at 100 ng/mL (100 μg/mL), The concentration of Recombinant Human MSP/MST1 that produces 50% optimal binding response is found to be 8-40 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA59872
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 4485
Molecular Weight (g/mol) M.W. (Observed): 81 kDa, 62 kDa and 30 kDa, reducing conditions; M.W. (theoretical): 78.5 kDa (MSP alpha/beta); 53.3 kDa (alpha chain), 25.2 kDa (beta chain)
Name MSP/MST1
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human MSP/MST1 protein Gln19-Gly711
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked heterodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol MST1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein MSP/MST1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.