missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MSP/MST1 (Cys672Ala) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
Description
Measured by its ability to activate MSP R/Ron in MDA-MB-453 or T47D human breast cancer cells. 80 ng/mL of the recombinant human MSP (C672A) significantly induces phosphorylation of MSP R/Ron measured by the Human Phospho-MSP R/Ron DuoSet™ IC Kit (Catalog # DYC1947 ). Measured by its binding ability in a functional ELISA. When Recombinant Human MSPR/Ron Protein (Catalog #1947-MS) is represent at 100 ng/mL (100 μg/mL), The concentration of Recombinant Human MSP/MST1 that produces 50% optimal binding response is found to be 4-32 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 4485 |
| Molecular Weight (g/mol) | 53 kDa ( alpha-chain), 25 kDa (beta-chain) |
| Name | MSP/MST1 (Cys672Ala) |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human MSP/MST1 protein Gln19-Arg483 (alpha) & Val484-Gly711 (Cys672Ala) (beta) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction